Heptyl acetoacetate ≥95.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAA0916-25MLEA
0
CNY
TCIAA0916-25ML
Heptyl acetoacetate ≥95.0% (通过GC)
Heptyl acetoacetate
分子式:
C₁₁H₂₀O₃ 分子量: 200,28 g/mol 沸点: 170°C |
MDL编号:
MFCD00027313 CAS号: 42598-96-9 EINECS: 255-906-0 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...