5-氨基乳清酸 ≥97.0% (通过总氮和滴定分析)

供应商: TCI EUROPE
Warning

Synonyms: 5-氨基-1,2,3,6-四氢-2,6-二氧代-4-嘧啶羧酸5-氨基-6-羧基-2,4-二羟基嘧啶

TCIAA0947-25GEA 0 CNY
TCIAA0947-25G
5-氨基乳清酸 ≥97.0% (通过总氮和滴定分析)
5-氨基乳清酸
分子式: C₅H₅N₃O₄
分子量: 171,11 g/mol
MDL编号: MFCD00010563
CAS号: 7164-43-4
EINECS: 230-514-2

现在订购

产品技术规格

Appearance White to Amber powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Solubility in Sodium hydroxide solution almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR