7-Acetoxy-4-bromomethylcoumarin ≥98.0% (通过HPLC,滴定分析) for HPLC labeling

供应商: TCI EUROPE
Danger

TCIAA0979-1GEA 0 CNY
TCIAA0979-1G
7-Acetoxy-4-bromomethylcoumarin ≥98.0% (通过HPLC,滴定分析) for HPLC labeling
7-Acetoxy-4-bromomethylcoumarin
分子式: C₁₂H₉BrO₄
MDL编号: MFCD00010707
CAS号: 2747-04-8
UN号: 3261
ADR: 8,II

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 182.0 to 186.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR