1-Acetylpyrrolidine ≥97.0% (通过GC)

供应商: TCI EUROPE

TCIAA1030-5GEA 0 CNY
TCIAA1030-5G
1-Acetylpyrrolidine ≥97.0% (通过GC)
1-Acetylpyrrolidine
分子式: C₆H₁₁NO
分子量: 113,16 g/mol
沸点: 225°C
MDL编号: MFCD00014096
CAS号: 4030-18-6

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR