Allyl-n-octyl ether ≥95.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAA1052-10MLEA
0
CNY
TCIAA1052-10ML
Allyl-n-octyl ether ≥95.0% (通过GC)
Allyl-n-octyl ether
分子式:
C₁₁H₂₂O 沸点: 206°C |
MDL编号:
MFCD00015294 CAS号: 3295-97-4 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...