4-Amidinobenzamide hydrochloride ≥92.0% (通过HPLC,总氮)

供应商: TCI EUROPE
Warning

TCIAA1062-10GEA 0 CNY
TCIAA1062-10G
4-Amidinobenzamide hydrochloride ≥92.0% (通过HPLC,总氮)
4-Amidinobenzamide hydrochloride
分子式: C₈H₉N₃O
分子量: 199,64 g/mol
MDL编号: MFCD00013011
CAS号: 59855-11-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 92.0 area%
Purity (with Total Nitrogen) min. 92.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR