Acetoxy-2-propanone ≥97.0% (通过GC) stabilised

供应商: TCI EUROPE


Stabilization: Stabilised with Mg₆Al₂(OH)₁₆CO₃·4H₂O

TCIAA1066-25MLEA 0 CNY
TCIAA1066-25ML
Acetoxy-2-propanone ≥97.0% (通过GC) stabilised
Acetoxy-2-propanone
分子式: C₅H₈O₃
分子量: 116,12 g/mol
沸点: 174°C
MDL编号: MFCD00042848
CAS号: 592-20-1
EINECS: 209-746-3

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR