3-乙酰基-2,5-二甲基呋喃 ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAA1121-25GEA
0
CNY
TCIAA1121-25G
3-乙酰基-2,5-二甲基呋喃 ≥98.0% (通过GC)
3-乙酰基-2,5-二甲基呋喃
分子式:
C₈H₁₀O₂ 分子量: 138,17 g/mol 沸点: 196°C 闪点: 82°C |
MDL编号:
MFCD00010281 CAS号: 10599-70-9 EINECS: 234-216-3 |
产品技术规格
Appearance | Colorless to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...