6-O-甲基鸟嘌呤 ≥97.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
Synonyms:
2-氨基-6-甲氧基嘌呤
, 6-甲氧基鸟嘌呤
TCIAA1151-100MGEA
0
CNY
TCIAA1151-100MG
6-O-甲基鸟嘌呤 ≥97.0% (通过滴定分析)
6-O-甲基鸟嘌呤
分子式:
C₆H₇N₅O 分子量: 165,16 g/mol |
MDL编号:
MFCD00133009 CAS号: 20535-83-5 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...