5-Methyl-3-heptyl acetate ≥95.0% (通过GC)

供应商: TCI EUROPE

TCIAA1379-5GEA 380 CNY
TCIAA1379-5G
5-Methyl-3-heptyl acetate ≥95.0% (通过GC)
5-Methyl-3-heptyl acetate
分子式: C₁₀H₂₀O₂
沸点: 185°C
MDL编号: MFCD00671490
CAS号: 375855-08-6

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR