1-Penten-3-ylacetate ≥97.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAA1542-5MLEA 0 CNY
TCIAA1542-5ML
1-Penten-3-ylacetate ≥97.0% (通过GC)
1-Penten-3-ylacetate
分子式: C₇H₁₂O₂
分子量: 128,17 g/mol
沸点: 132°C
MDL编号: MFCD02093427
CAS号: 10500-11-5
UN号: 3272
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR