3-氨基-5-硝基苯并异噻唑 ≥93.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAA1562-5GEA 0 CNY
TCIAA1562-5G
3-氨基-5-硝基苯并异噻唑 ≥93.0% (通过滴定分析)
3-氨基-5-硝基苯并异噻唑
分子式: C₇H₅N₃O₂S
分子量: 195,20 g/mol
MDL编号: MFCD00270828
CAS号: 84387-89-3

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Nonaqueous Titration) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR