4-Phenylbutylamine hydrochloride ≥98.0% (通过HPLC,总氮)

供应商: TCI EUROPE
Warning

TCIAA1590-25GEA 0 CNY
TCIAA1590-25G
4-Phenylbutylamine hydrochloride ≥98.0% (通过HPLC,总氮)
4-Phenylbutylamine hydrochloride
分子式: C₁₀H₁₅N
分子量: 185,70 g/mol
MDL编号: MFCD03093616
CAS号: 30684-06-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 163.0 to 167.0 deg-C
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR