(1S,2S)-2-(Anthracene-2,3-dicarboximido)cyclohexanecarboxylic acid ≥97.0% (通过HPLC)
供应商: TCI EUROPE
|
Warning
|
TCIAA1658-100MGEA
0
CNY
TCIAA1658-100MG
(1S,2S)-2-(Anthracene-2,3-dicarboximido)cyclohexanecarboxylic acid ≥97.0% (通过HPLC)
(1S,2S)-2-(Anthracene-2,3-dicarboximido)cyclohexanecarboxylic acid
分子式:
C₂₃H₁₉NO₄ 分子量: 373,41 g/mol |
CAS号:
446044-45-7 |
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Specific rotation [a]20/D | +38.0 to +45.0 deg (C=0.2,THF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...