4-Amino-2-nitrobenzoic acid ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAA1686-1GEA 0 CNY
TCIAA1686-1G
4-Amino-2-nitrobenzoic acid ≥98.0% (通过HPLC,滴定分析)
4-Amino-2-nitrobenzoic acid
分子式: C₇H₆N₂O₄
分子量: 182,14 g/mol
溶点: 239°C
MDL编号: MFCD00189376
CAS号: 610-36-6

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR