4,7,8,9-Tetra-O-acetyl-N-acetylneuraminic acid methyl ester ≥90.0% (通过HPLC)
供应商: TCI EUROPE
TCIAA1822-1GEA
0
CNY
TCIAA1822-1G
4,7,8,9-Tetra-O-acetyl-N-acetylneuraminic acid methyl ester ≥90.0% (通过HPLC)
4,7,8,9-Tetra-O-acetyl-N-acetylneuraminic acid methyl ester
分子式:
C₂₀H₂₉NO₁₃ 分子量: 491,45 g/mol 存储条件: Freezer |
MDL编号:
MFCD08276266 CAS号: 84380-10-9 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 90.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...