9-Azajulolidine ≥97.0% (通过GC)
供应商: TCI EUROPE
TCIAA1854-1GEA
0
CNY
TCIAA1854-1G
9-Azajulolidine ≥97.0% (通过GC)
9-Azajulolidine
分子式:
C₁₁H₁₄N₂ 分子量: 174,25 g/mol 沸点: 145°C 存储条件: Refrigerator |
CAS号:
6052-72-8 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 70.0 to 74.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...