3-(2-Aminoethylamino)propyltriethoxysilane ≥96.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAA1923-25GEA 0 CNY
TCIAA1923-25G
3-(2-Aminoethylamino)propyltriethoxysilane ≥96.0% (通过GC)
3-(2-Aminoethylamino)propyltriethoxysilane
分子式: C₁₁H₂₈N₂O₃Si
分子量: 264,44 g/mol
沸点: 156°C
MDL编号: MFCD00138097
CAS号: 5089-72-5
EINECS: 225-806-1

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR