3-Acetamidopiperidine ≥99.0% (通过GC,滴定分析)

供应商: TCI EUROPE

TCIAA1934-5GEA 0 CNY
TCIAA1934-5G
3-Acetamidopiperidine ≥99.0% (通过GC,滴定分析)
3-Acetamidopiperidine
分子式: C₇H₁₄N₂O
分子量: 142,20 g/mol
CAS号: 5810-55-9

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 79.0 to 83.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR