1-Acetoxyindan ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAA1975-5GEA 0 CNY
TCIAA1975-5G
1-Acetoxyindan ≥98.0% (通过GC)
1-Acetoxyindan
分子式: C₁₁H₁₂O₂
分子量: 176,22 g/mol
沸点: 241°C
MDL编号: MFCD00053467
CAS号: 26452-98-2

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR