4-Amino-2,5-dimethoxy-N-phenylsulphonamide ≥98.0% (通过HPLC,滴定分析)
供应商: TCI EUROPE
TCIAA1987-25GEA
0
CNY
TCIAA1987-25G
4-Amino-2,5-dimethoxy-N-phenylsulphonamide ≥98.0% (通过HPLC,滴定分析)
4-Amino-2,5-dimethoxy-N-phenylsulphonamide
分子式:
C₁₄H₁₆N₂O₄S 分子量: 308,35 g/mol |
MDL编号:
MFCD00025753 CAS号: 52298-44-9 EINECS: 257-821-4 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 184.0 to 188.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...