4-氨基-4′-硝基二苯硫醚 ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

Synonyms: 4-氨基-4′-硝基二苯基硫醚

TCIAA2077-25GEA 0 CNY
TCIAA2077-25G
4-氨基-4′-硝基二苯硫醚 ≥98.0% (通过GC,滴定分析)
4-氨基-4′-硝基二苯硫醚
分子式: C₁₂H₁₀N₂O₂S
分子量: 246,28 g/mol
MDL编号: MFCD00007881
CAS号: 101-59-7
EINECS: 202-957-1

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 141.0 to 145.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR