4-Anilinopyridine ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAA2087-5GEA 1275 CNY
TCIAA2087-5G
4-Anilinopyridine ≥98.0% (通过GC,滴定分析)
4-Anilinopyridine
分子式: C₁₁H₁₀N₂
分子量: 170,22 g/mol
MDL编号: MFCD00457758
CAS号: 22961-45-1
EINECS: 245-352-8

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystalline
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 175.0 to 179.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR