4-Amino-5-fluoro-2-methoxypyrimidine ≥99.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAA2125-5GEA 0 CNY
TCIAA2125-5G
4-Amino-5-fluoro-2-methoxypyrimidine ≥99.0% (通过滴定分析)
4-Amino-5-fluoro-2-methoxypyrimidine
分子式: C₅H₆FN₃O
分子量: 143,12 g/mol
MDL编号: MFCD09264289
CAS号: 1993-63-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 191.0 to 195.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR