5-Acetamido-2-nitrobenzoic acid ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAA2156-5GEA 900 CNY
TCIAA2156-5G
5-Acetamido-2-nitrobenzoic acid ≥98.0% (通过滴定分析)
5-Acetamido-2-nitrobenzoic acid
分子式: C₉H₈N₂O₅
溶点: 225°C
MDL编号: MFCD00035889
CAS号: 4368-83-6
EINECS: 224-461-4

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR