5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide ≥97.0% (以总氮计)
供应商: TCI EUROPE
|
Warning
|
TCIAA2169-25GEA
0
CNY
TCIAA2169-25G
5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide ≥97.0% (以总氮计)
5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide
分子式:
C₁₆H₂₀I₃N₃O₇ 分子量: 747,06 g/mol 溶点: 275°C |
MDL编号:
MFCD08063354 CAS号: 31127-80-7 EINECS: 445-830-8 |
产品技术规格
Appearance | White to Light red to Green powder to crystalline |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...