1-Acenaphthenone ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAA2185-1GEA 0 CNY
TCIAA2185-1G
1-Acenaphthenone ≥98.0% (通过GC)
1-Acenaphthenone
分子式: C₁₂H₈O
分子量: 168,20 g/mol
沸点: 180°C
MDL编号: MFCD00156651
CAS号: 2235-15-6

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystalline
Purity (GC) min. 98.0 %
Melting point 120.0 to 124.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR