O-烯丙基-2,2,2-三氯乙酰亚胺酯 ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Danger
|
Synonyms:
2,2,2-三氯亚氨逐乙酸烯丙酯
TCIAA2186-5GEA
0
CNY
TCIAA2186-5G
O-烯丙基-2,2,2-三氯乙酰亚胺酯 ≥98.0% (通过GC)
O-烯丙基-2,2,2-三氯乙酰亚胺酯
分子式:
C₅H₆Cl₃NO 分子量: 202,46 g/mol 沸点: 74°C 闪点: 74°C 存储条件: Refrigerator |
MDL编号:
MFCD00190710 CAS号: 51479-73-3 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...