2-Chloroaniline-5-sulphonic acid ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE

TCIAA2196-25GEA 540 CNY
TCIAA2196-25G
2-Chloroaniline-5-sulphonic acid ≥98.0% (通过HPLC,滴定分析)
2-Chloroaniline-5-sulphonic acid
分子式: C₆H₆ClNO₃S
MDL编号: MFCD00035764
CAS号: 98-36-2
EINECS: 202-661-2

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Water max. 9.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR