1-(2-Amino-1-naphthyl)isoquinoline ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAA2221-1GEA
0
CNY
TCIAA2221-1G
1-(2-Amino-1-naphthyl)isoquinoline ≥98.0% (通过滴定分析)
1-(2-Amino-1-naphthyl)isoquinoline
分子式:
C₁₉H₁₄N₂ 分子量: 270,34 g/mol |
CAS号:
664302-70-9 |
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...