(1S,3R)-3-Aminocyclohexanecarboxylic acid ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE

TCIAA2286-200MGEA 0 CNY
TCIAA2286-200MG
(1S,3R)-3-Aminocyclohexanecarboxylic acid ≥98.0% (通过GC,滴定分析)
(1S,3R)-3-Aminocyclohexanecarboxylic acid
分子式: C₇H₁₃NO₂
分子量: 143,19 g/mol
溶点: 240°C
MDL编号: MFCD09753648
CAS号: 81131-40-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +3.0 to +6.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR