Allyl(tert-butyl)dimethylsilane ≥97.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAA2298-5GEA 0 CNY
TCIAA2298-5G
Allyl(tert-butyl)dimethylsilane ≥97.0% (通过GC)
Allyl(tert-butyl)dimethylsilane
分子式: C₉H₂₀Si
分子量: 156,34 g/mol
沸点: 152°C
MDL编号: MFCD18207712
CAS号: 74472-22-3
UN号: 1993
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR