(R)-(+)-2′-氨基-1,1′-联萘-2-醇 ≥98.0% (通过HPLC)

供应商: TCI EUROPE
Danger

Synonyms: (R)-NOBIN(R)-(+)-2-氨基-2'-羟基-1,1'-联萘

TCIAA2316-100MGEA 0 CNY
TCIAA2316-100MG
(R)-(+)-2′-氨基-1,1′-联萘-2-醇 ≥98.0% (通过HPLC)
(R)-(+)-2′-氨基-1,1′-联萘-2-醇
分子式: C₂₀H₁₅NO
分子量: 285,35 g/mol
MDL编号: MFCD01882346
CAS号: 137848-28-3

现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR