1-(2-Aminoethyl)-4-methylpiperazine hydrochloride ≥98.0% (通过总氮和滴定分析)

供应商: TCI EUROPE
Warning

TCIAA2398-1GEA 1200 CNY
TCIAA2398-1G
1-(2-Aminoethyl)-4-methylpiperazine hydrochloride ≥98.0% (通过总氮和滴定分析)
1-(2-Aminoethyl)-4-methylpiperazine hydrochloride
分子式: C₇H₁₇N₃
CAS号: 646051-17-4

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR