2-(Methylsulphonyl)ethanamine hydrochloride ≥98.0% (通过总氮和滴定分析)

供应商: TCI EUROPE

TCIAA2464-5GEA 575 CNY
TCIAA2464-5G
2-(Methylsulphonyl)ethanamine hydrochloride ≥98.0% (通过总氮和滴定分析)
2-(Methylsulphonyl)ethanamine hydrochloride
分子式: C₃H₁₀ClNO₂S
MDL编号: MFCD03840162
CAS号: 104458-24-4

现在订购

产品技术规格

Appearance White to Light yellow powder to crystalline
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 168.0 to 172.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR