(S)-(+)-DBD-APy [=(S)-(+)-4-(N,N-Dimethylaminosulphonyl)-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole] ≥98.0% (通过HPLC) HPLC labeling reagent for e.e. determination

供应商: TCI EUROPE
Warning

TCIAA5560-100MGEA 0 CNY
TCIAA5560-100MG
(S)-(+)-DBD-APy [=(S)-(+)-4-(N,N-Dimethylaminosulphonyl)-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole] ≥98.0% (通过HPLC) HPLC labeling reagent for e.e. determination
(S)-(+)-DBD-APy [=(S)-(+)-4-(N,N-Dimethylaminosulphonyl)-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole]
分子式: C₁₂H₁₇N₅O₃S
分子量: 311,36 g/mol
MDL编号: MFCD00191378
CAS号: 143112-50-9

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 99.0 ee%
Melting point 128.0 to 132.0 deg-C
Specific rotation [a]20/D +3.0 to +7.0 deg (C=1, CH3CN)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR