(R)-(-)-DBD-Py-NCS [=(R)-(-)-4-(N,N-Dimethylaminosulphonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)-2,1,3-benzoxadiazole] ≥98.0% (通过HPLC) for HPLC labeling

供应商: TCI EUROPE

TCIAA5568-100MGEA 0 CNY
TCIAA5568-100MG
(R)-(-)-DBD-Py-NCS [=(R)-(-)-4-(N,N-Dimethylaminosulphonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)-2,1,3-benzoxadiazole] ≥98.0% (通过HPLC) for HPLC labeling
(R)-(-)-DBD-Py-NCS [=(R)-(-)-4-(N,N-Dimethylaminosulphonyl)-7-(3-isothiocyanatopyrrolidin-1-yl)-2,1,3-benzoxadiazole]
分子式: C₁₃H₁₅N₅O₃S₂
分子量: 353,42 g/mol
存储条件: Refrigerator
MDL编号: MFCD00671493
CAS号: 163927-31-9

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 99.0 ee%
Specific rotation [a]20/D -280.0 to -290.0 deg (C=0.4, CH3CN)
Effect test of HPLC derivatization Effective as labeling agent for DL-1-Phenylethylamine

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR