(S)-(+)-NBD-Py-NCS [=(S)-(+)-4-(3-Isothiocyanatopyrrolidin-1-yl)-7-nitro-2,1,3-benzoxadiazole] ≥98.0% (通过HPLC) HPLC labeling reagent for e.e. determination

供应商: TCI EUROPE
Warning

TCIAA5578-100MGEA 0 CNY
TCIAA5578-100MG
(S)-(+)-NBD-Py-NCS [=(S)-(+)-4-(3-Isothiocyanatopyrrolidin-1-yl)-7-nitro-2,1,3-benzoxadiazole] ≥98.0% (通过HPLC) HPLC labeling reagent for e.e. determination
(S)-(+)-NBD-Py-NCS [=(S)-(+)-4-(3-Isothiocyanatopyrrolidin-1-yl)-7-nitro-2,1,3-benzoxadiazole]
分子式: C₁₁H₉N₅O₃S
分子量: 291,29 g/mol
存储条件: Refrigerator
MDL编号: MFCD02093435
CAS号: 163927-30-8

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 99.0 ee%
Specific rotation [a]20/D +530 to +600 deg (C=0.2, CH3CN)
Effect test of HPLC derivatization Effective as labeling agent for DL-Valine, DL-Phenylalanine

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR