Benzanthrone ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAB0019-500GEA
0
CNY
TCIAB0019-500G
TCIAB0019-25G
Benzanthrone ≥98.0% (通过GC)
Benzanthrone
分子式:
C₁₇H₁₀O 分子量: 230,27 g/mol |
MDL编号:
MFCD00003585 CAS号: 82-05-3 EINECS: 201-393-3 |
产品技术规格
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 172.0 to 176.0 deg-C |
Solubility in hot Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...