2-溴乙基苯甲酸酯 ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAB0065-25MLEA 0 CNY
TCIAB0065-25ML
2-溴乙基苯甲酸酯 ≥98.0% (通过GC)
2-溴乙基苯甲酸酯
分子式: C₉H₉BrO₂
分子量: 229,07 g/mol
沸点: 285°C
MDL编号: MFCD00059462
CAS号: 939-54-8

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR