2-Chloroethyl benzoate ≥99.0% (通过GC)

供应商: TCI EUROPE

TCIAB0068-25MLEA 0 CNY
TCIAB0068-25ML
2-Chloroethyl benzoate ≥99.0% (通过GC)
2-Chloroethyl benzoate
分子式: C₉H₉ClO₂
分子量: 184,62 g/mol
沸点: 257°C
MDL编号: MFCD00045290
CAS号: 939-55-9

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR