3,5-Diphenyl-1-(m-tolyl)formazan ≥98.0% (通过HPLC)

供应商: TCI EUROPE

TCIAB0283-100MGEA 0 CNY
TCIAB0283-100MG
3,5-Diphenyl-1-(m-tolyl)formazan ≥98.0% (通过HPLC)
3,5-Diphenyl-1-(m-tolyl)formazan
分子式: C₂₀H₁₈N₄
分子量: 314,39 g/mol
MDL编号: MFCD00059314
CAS号: 71274-65-2

现在订购

产品技术规格

Appearance Orange to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 154.0 to 158.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR