Benzoyl-DL-methionine ≥99.0% (通过滴定分析)

供应商: TCI EUROPE

Synonyms: Bz-DL-Met-OH2-Benzamido-4-(methylthio)butanoic acid

TCIAB0300-1GEA 0 CNY
TCIAB0300-1G
Benzoyl-DL-methionine ≥99.0% (通过滴定分析)
Benzoyl-DL-methionine
分子式: C₁₂H₁₅NO₃S
分子量: 253,32 g/mol
MDL编号: MFCD00026044
CAS号: 4703-38-2

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 99.0 %
Melting point 150.0 to 154.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR