Benzoyl-DL-phenylalanine ≥98.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAB0302-10GEA 0 CNY
TCIAB0302-10G
Benzoyl-DL-phenylalanine ≥98.0% (通过滴定分析)
Benzoyl-DL-phenylalanine
分子式: C₁₆H₁₅NO₃
分子量: 269,30 g/mol
MDL编号: MFCD00037252
CAS号: 2901-76-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 184.0 to 188.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR