2,3-Diphenyl-5-methyltetrazolium chloride ≥95.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAB0313-100MGEA 0 CNY
TCIAB0313-100MG
2,3-Diphenyl-5-methyltetrazolium chloride ≥95.0% (通过滴定分析)
2,3-Diphenyl-5-methyltetrazolium chloride
分子式: C₁₄H₁₃ClN₄
MDL编号: MFCD00060025
CAS号: 6275-01-0

现在订购

产品技术规格

Purity (HPLC) min. 94.0 area%
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR