2,3-Diphenyl-5-(4-chlorophenyl)tetrazolium chloride ≥92.0% (通过滴定分析)
供应商: TCI EUROPE
TCIAB0329-100MGEA
0
CNY
TCIAB0329-100MG
2,3-Diphenyl-5-(4-chlorophenyl)tetrazolium chloride ≥92.0% (通过滴定分析)
2,3-Diphenyl-5-(4-chlorophenyl)tetrazolium chloride
分子式:
C₁₉H₁₄Cl₂N₄ 溶点: 255°C |
MDL编号:
MFCD00060031 CAS号: 10557-51-4 |
产品技术规格
Purity (Nonaqueous Titration) | min. 92.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...