2-(2-Benzothiazolyl)-3,5-diphenyltetrazolium bromide ≥95.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAB0333-100MGEA 0 CNY
TCIAB0333-100MG
2-(2-Benzothiazolyl)-3,5-diphenyltetrazolium bromide ≥95.0% (通过滴定分析)
2-(2-Benzothiazolyl)-3,5-diphenyltetrazolium bromide
分子式: C₂₀H₁₄BrN₅S
MDL编号: MFCD00135111
CAS号: 55699-64-4

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (Bromination Method) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR