4-Ethoxyphenyl-4-butylbenzoate ≥99.0% (通过GC)

供应商: TCI EUROPE

TCIAB0375-1GEA 0 CNY
TCIAB0375-1G
4-Ethoxyphenyl-4-butylbenzoate ≥99.0% (通过GC)
4-Ethoxyphenyl-4-butylbenzoate
分子式: C₁₉H₂₂O₃
分子量: 298,38 g/mol
MDL编号: MFCD00059426
CAS号: 62716-65-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 59.0 to 63.0 deg-C
Solubility in Methanol within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR