Biurea

供应商: TCI EUROPE

TCIAB0512-25GEA 1825 CNY
TCIAB0512-5G TCIAB0512-25G
Biurea
Biurea
分子式: C₂H₆N₄O₂
分子量: 118,10 g/mol
MDL编号: MFCD00025398
CAS号: 110-21-4
EINECS: 203-747-2

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Solubility in hot Water almost transparency
Elemental analysis (Nitrogen) 45.00 to 48.00 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR