2-溴辛酸 ≥97.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Danger

Synonyms: 2-Bromocaprylic acid

TCIAB0627-25GEA 0 CNY
TCIAB0627-25G
2-溴辛酸 ≥97.0% (通过GC,滴定分析)
2-溴辛酸
分子式: C₈H₁₅BrO₂
分子量: 223,11 g/mol
沸点: 164°C
MDL编号: MFCD00004219
CAS号: 2623-82-7
EINECS: 220-079-7
UN号: 3265
ADR: 8,II

现在订购

产品技术规格

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR